Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286609.1 | NP_001273538.1 | 4 | 668 | ||
NM_032131.5 | NP_115507.4 | 5 | 741 | |||
XM_005267154.3 | XP_005267211.1 | 5 | 1022 | |||
XM_005267155.2 | XP_005267212.1 | 5 | 757 | |||
XM_005267157.3 | XP_005267214.1 | 5 | 665 | |||
XM_006715573.2 | XP_006715636.1 | 6 | 847 | |||
XM_006715574.2 | XP_006715637.1 | 2 | 177 | |||
XM_011536166.1 | XP_011534468.1 | 6 | 832 | |||
XM_011536167.1 | XP_011534469.1 | 5 | 735 | |||
XM_011536168.2 | XP_011534470.1 | 5 | 735 | |||
XM_011536170.2 | XP_011534472.1 | 5 | 735 | |||
XM_011536172.2 | XP_011534474.1 | 4 | 592 | |||
XM_011536176.2 | XP_011534478.1 | 5 | 735 | |||
XM_017011350.1 | XP_016866839.1 | 5 | 747 | |||
XM_017011351.1 | XP_016866840.1 | 5 | 735 | |||
XM_017011352.1 | XP_016866841.1 | 5 | 747 | |||
GenBank mRNA | AK095234.1 | - | 4 | 584 | ||
AK292499.1 | - | 5 | 735 | |||
AK294574.1 | - | 4 | 668 | |||
AL136754.1 | - | 1 | 91 | |||
BC030603.2 | - | 5 | 735 | 4824783 |