Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032135.3 | NP_115511.3 | 1 | 393 | ||
GenBank mRNA | AK124110.1 | - | 1 | 383 | ||
AK302615.1 | - | 1 | 347 | |||
BC039878.1 | - | 1 | 352 | 5163160 | ||
CR933626.1 | - | 1 | 350 | |||
DB036759.1 | - | 1 | 393 |