Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032151.4 | NP_115527.3 | 2 | 149 | ||
GenBank mRNA | AF499009.1 | - | 2 | 69 | ||
AK098495.1 | - | 2 | 121 | |||
AL136721.1 | - | 2 | 149 | |||
BC054021.1 | - | 2 | 152 | 6385036 |