Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032169.4 | NP_115545.3 | 9 | 1520 | ||
NR_132426.1 | 9 | 1520 | ||||
NR_132427.1 | 9 | 1520 | ||||
NR_132428.1 | 9 | 1520 | ||||
GenBank mRNA | AJ608287.1 | - | 9 | 1471 | ||
AK022654.1 | - | 2 | 114 | |||
AK131265.1 | - | 11 | 1317 | |||
AK298627.1 | - | 1 | 327 | |||
AK308380.1 | - | 10 | 1110 | |||
AL832873.1 | - | 9 | 1471 | |||
AL833721.1 | - | 9 | 1038 | |||
AX803783.1 | - | 9 | 1377 | |||
AX803785.1 | - | 9 | 1140 | |||
BC019607.1 | - | 4 | 416 | |||
BC125204.1 | - | 9 | 1455 | 40123454 | ||
BC125205.1 | - | 9 | 1455 |
Gene Symbol: NPHP3-ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037804.1 | 34 | 4981 | |||
GenBank mRNA | AB082531.1 | - | 34 | 4981 |