Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032206.4 | NP_115582.4 | 41 | 5093 | ||
XM_005256193.2 | XP_005256250.1 | 42 | 5154 | |||
XM_005256194.3 | XP_005256251.1 | 42 | 5365 | |||
XM_005256195.2 | XP_005256252.1 | 42 | 5151 | |||
XM_005256196.2 | XP_005256253.1 | 41 | 5070 | |||
XM_005256197.2 | XP_005256254.1 | 41 | 5067 | |||
XM_006721297.2 | XP_006721360.1 | 42 | 5142 | |||
XM_006721298.2 | XP_006721361.1 | 41 | 5064 | |||
XM_011523373.2 | XP_011521675.1 | 42 | 5301 | |||
XM_011523376.2 | XP_011521678.1 | 37 | 2980 | |||
XM_017023766.1 | XP_016879255.1 | 41 | 5070 | |||
XM_017023767.1 | XP_016879256.1 | 41 | 5055 | |||
XM_017023768.1 | XP_016879257.1 | 41 | 5067 | |||
GenBank mRNA | AF389420.1 | - | 40 | 4966 | ||
AK027414.1 | - | 21 | 1683 | |||
AK027416.1 | - | 21 | 1682 | |||
AK074133.1 | - | 30 | 2483 | |||
AK090439.1 | - | 37 | 4206 | |||
BC050527.1 | - | 26 | 1986 | |||
BC063566.1 | - | 24 | 1879 | |||
EF452236.1 | - | 40 | 4966 |