Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032233.2 | NP_115609.2 | 4 | 499 | ||
NM_199123.1 | NP_954574.1 | 4 | 499 | |||
XM_011537231.2 | XP_011535533.2 | 4 | 451 | |||
XM_011537232.2 | XP_011535534.1 | 4 | 1629 | |||
XM_011537233.2 | XP_011535535.1 | 4 | 451 | |||
XM_011537235.1 | XP_011535537.1 | 4 | 451 | |||
XM_017021699.1 | XP_016877188.1 | 3 | 347 | |||
XM_017021700.1 | XP_016877189.1 | 4 | 535 | |||
XM_017021701.1 | XP_016877190.1 | 4 | 501 | |||
GenBank mRNA | AK026680.1 | - | 3 | 253 | ||
AK302882.1 | - | 4 | 1842 | |||
BC009054.1 | - | 1 | 125 | |||
BC066931.1 | - | 4 | 400 | |||
BC120967.1 | - | 4 | 354 | |||
BC120968.2 | - | 4 | 354 | |||
BC127624.1 | - | 4 | 354 | |||
BC127625.1 | - | 4 | 354 | |||
BC142995.1 | - | 4 | 511 | 8860450 | ||
BC148251.1 | - | 3 | 350 | |||
BX161441.1 | - | 4 | 398 | |||
BX161471.1 | - | 4 | 489 | |||
CB988965.1 | - | 4 | 526 |