Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286212.1 | NP_001273141.1 | 6 | 1070 | ||
NM_001286213.1 | NP_001273142.1 | 6 | 1037 | |||
NM_032256.2 | NP_115632.1 | 7 | 1170 | |||
XM_011538831.2 | XP_011537133.1 | 7 | 1179 | |||
XM_011538832.2 | XP_011537134.1 | 7 | 854 | |||
XM_011538833.2 | XP_011537135.1 | 6 | 671 | |||
XM_017020022.1 | XP_016875511.1 | 7 | 761 | |||
XM_017020023.1 | XP_016875512.1 | 8 | 1002 | |||
GenBank mRNA | AK057287.1 | - | 7 | 889 | ||
AK097511.1 | - | 2 | 115 | |||
AL136855.1 | - | 7 | 872 | |||
BC030284.1 | - | 6 | 778 | 5240522 | ||
BC060798.1 | - | 7 | 933 | 30348706 | ||
DA007410.1 | - | 6 | 478 |