Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032270.4 | NP_115646.2 | 3 | 1728 | ||
XM_006710960.3 | XP_006711023.1 | 3 | 3370 | |||
XM_011542282.2 | XP_011540584.1 | 3 | 1715 | |||
XM_011542283.2 | XP_011540585.1 | 4 | 3282 | |||
XM_017002503.1 | XP_016857992.1 | 3 | 1575 | |||
XM_017002504.1 | XP_016857993.1 | 4 | 1484 | |||
GenBank mRNA | AB081509.1 | - | 3 | 1609 | ||
AK127881.1 | - | 3 | 1665 | |||
AL136919.1 | - | 1 | 1228 | |||
BC113973.1 | - | 3 | 1535 | 40007964 |