Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032271.2 | NP_115647.2 | 19 | 1943 | ||
XM_005255627.4 | XP_005255684.1 | 19 | 1983 | |||
XM_011522700.1 | XP_011521002.1 | 19 | 1985 | |||
GenBank mRNA | AK090624.1 | - | 19 | 1957 | ||
AL136921.1 | - | 18 | 1839 | |||
AY569455.1 | - | 18 | 1896 | |||
BC024267.1 | - | 17 | 1729 |