Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318739.1 | NP_001305668.1 | 2 | 564 | ||
NM_001318740.1 | NP_001305669.1 | 2 | 461 | |||
NM_001318741.1 | NP_001305670.1 | 2 | 564 | |||
NM_032299.3 | NP_115675.1 | 2 | 461 | |||
XM_017018404.1 | XP_016873893.1 | 2 | 436 | |||
GenBank mRNA | AK056993.1 | - | 2 | 372 | ||
AK298227.1 | - | 2 | 526 | |||
AY364246.1 | - | 2 | 118 | |||
BC004169.2 | - | 2 | 386 | 2821548 | ||
BC021074.1 | - | 2 | 195 | |||
BM723994.1 | - | 2 | 210 | |||
CN261961.1 | - | 2 | 394 | |||
DB039638.1 | - | 2 | 461 |