Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318739.1 | NP_001305668.1 | 8 | 1125 | ||
NM_001318740.1 | NP_001305669.1 | 7 | 930 | |||
NM_001318741.1 | NP_001305670.1 | 7 | 1033 | |||
NM_032299.3 | NP_115675.1 | 8 | 1022 | |||
GenBank mRNA | AK056993.1 | - | 8 | 933 | ||
AK298227.1 | - | 8 | 1087 | |||
BC004169.2 | - | 8 | 947 | 2821548 | ||
BC021074.1 | - | 8 | 756 | |||
BM723994.1 | - | 7 | 679 |