Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032303.4 | NP_115679.2 | 4, 5 | 642 | ||
NR_036651.1 | 3, 4 | 478 | ||||
XM_011519091.2 | XP_011517393.1 | 4, 5 | 642 | |||
GenBank mRNA | AK098721.1 | - | 4, 5 | 613 | ||
AK289929.1 | - | 4, 5 | 470 | |||
AK303645.1 | - | 3, 4 | 284 | |||
AL833735.1 | - | 2, 3 | 156 | |||
AY093428.1 | - | 4, 5 | 481 | |||
BC095451.1 | - | 4, 5 | 419 | 30405073 |