Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033002.3 | NP_001028174.2 | 4 | 973 | ||
NM_001160243.1 | NP_001153715.1 | 4 | 973 | |||
NM_001160244.1 | NP_001153716.1 | 4 | 973 | |||
NM_001160266.1 | NP_001153738.1 | 4 | 973 | |||
NR_027682.1 | 4 | 973 | ||||
GenBank mRNA | AA745783.1 | - | 2 | 359 | ||
AK295394.1 | - | 4 | 414 | |||
AK300409.1 | - | 4 | 429 | |||
AY680654.1 | - | 4 | 403 | |||
AY680655.1 | - | 4 | 403 | |||
AY680656.1 | - | 4 | 403 | |||
AY775314.1 | - | 4 | 973 | |||
AY775315.1 | - | 4 | 973 | |||
AY775316.1 | - | 4 | 973 | |||
AY775317.1 | - | 4 | 973 | |||
AY775318.1 | - | 4 | 973 | |||
BC004451.1 | - | 4 | 416 | 2820882 | ||
BC013831.1 | - | 4 | 444 | |||
BC051849.2 | - | 4 | 406 | 5493799 | ||
N41653.1 | - | 4 | 371 |