Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032320.6 | NP_115696.2 | 2 | 332 | ||
XM_017018406.1 | XP_016873895.1 | 2 | 322 | |||
XM_017018407.1 | XP_016873896.1 | 2 | 302 | |||
XM_017018408.1 | XP_016873897.1 | 2 | 302 | |||
GenBank mRNA | AF329102.2 | - | 2 | 332 | ||
AK095946.1 | - | 2 | 261 | |||
AK292392.1 | - | 2 | 257 | |||
AL832981.1 | - | 2 | 313 | |||
AY221959.2 | - | 2 | 375 | |||
BC005071.1 | - | 2 | 294 | 3531347 |