Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTPCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF416713.1 | - | 2 | 142 | ||
AK316591.1 | - | 2 | 232 | |||
BC005102.1 | - | 2 | 180 | 3619026 | ||
BG473925.1 | - | 2 | 351 | |||
HY133356.1 | - | 2 | 274 |