Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTPCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA635564.1 | - | 2 | 290 | ||
AF416713.1 | - | 3 | 278 | |||
AK311229.1 | - | 2 | 204 | |||
AK316591.1 | - | 3 | 368 | |||
BC005102.1 | - | 3 | 316 | 3619026 | ||
BG106026.1 | - | 2 | 151 | |||
BG473925.1 | - | 3 | 487 | |||
BQ001389.1 | - | 2 | 719 | |||
DC332692.1 | - | 2 | 205 | |||
HY133356.1 | - | 3 | 410 | |||
W80669.1 | - | 2 | 136 |