Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297423.1 | NP_001284352.1 | 10 | 1382 | ||
NM_001297424.1 | NP_001284353.1 | 8 | 698 | |||
NM_001297425.1 | NP_001284354.1 | 8 | 695 | |||
NM_001297426.1 | NP_001284355.1 | 11 | 1162 | |||
NM_001297427.1 | NP_001284356.1 | 10 | 1418 | |||
NM_001297428.1 | NP_001284357.1 | 8 | 734 | |||
NM_032326.3 | NP_115702.1 | 10 | 921 | |||
XM_005272301.2 | XP_005272358.1 | 9 | 812 | |||
XM_005272303.4 | XP_005272360.1 | 11 | 1078 | |||
XM_005272304.1 | XP_005272361.1 | 10 | 887 | |||
XM_005272307.4 | XP_005272364.1 | 10 | 847 | |||
XM_011513576.1 | XP_011511878.1 | 6 | 439 | |||
XM_017008701.1 | XP_016864190.1 | 11 | 998 | |||
XM_017008702.1 | XP_016864191.1 | 8 | 645 | |||
XM_017008703.1 | XP_016864192.1 | 11 | 1440 | |||
XM_017008704.1 | XP_016864193.1 | 9 | 1253 | |||
XM_017008705.1 | XP_016864194.1 | 7 | 590 | |||
GenBank mRNA | AK090848.1 | - | 10 | 1312 | ||
AK309132.1 | - | 10 | 847 | |||
AL834199.1 | - | 11 | 1032 | |||
BC005158.2 | - | 10 | 834 | 3505819 | ||
BI819231.1 | - | 8 | 642 | |||
CB992640.1 | - | 8 | 662 | |||
CF242841.1 | - | 8 | 637 |