Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM213A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243778.1 | NP_001230707.1 | 5 | 504 | ||
NM_001243779.1 | NP_001230708.1 | 5 | 574 | |||
NM_001243780.1 | NP_001230709.1 | 5 | 1070 | |||
NM_001243781.1 | NP_001230710.1 | 5 | 587 | |||
NM_001243782.1 | NP_001230711.1 | 5 | 487 | |||
NM_032333.4 | NP_115709.3 | 5 | 604 | |||
XM_011540265.2 | XP_011538567.1 | 5 | 907 | |||
XM_011540266.2 | XP_011538568.1 | 5 | 1310 | |||
XM_017016775.1 | XP_016872264.1 | 5 | 591 | |||
GenBank mRNA | AF305824.1 | - | 3 | 260 | ||
AK074643.1 | - | 5 | 549 | |||
AK075447.1 | - | 5 | 579 | |||
AK225387.1 | - | 5 | 478 | |||
AK315440.1 | - | 5 | 498 | |||
AY359059.1 | - | 5 | 497 | |||
BC005871.2 | - | 5 | 466 | 3010078 | ||
BE742158.1 | - | 5 | 574 | |||
CN396006.1 | - | 5 | 456 | |||
DA136257.1 | - | 5 | 488 |