Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRMS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032352.3 | NP_115728.2 | 3, 4 | 481 | ||
XM_005268128.1 | XP_005268185.1 | 3, 4 | 554 | |||
XM_006720274.2 | XP_006720337.1 | 4, 5 | 544 | |||
XM_006720275.2 | XP_006720338.1 | 4, 5 | 544 | |||
XM_017021703.1 | XP_016877192.1 | 3, 4 | 301 | |||
XM_017021704.1 | XP_016877193.1 | 4, 5 | 455 | |||
XM_017021705.1 | XP_016877194.1 | 3, 4 | 378 | |||
GenBank mRNA | AK096496.1 | - | 2, 3 | 204 | ||
AK297106.1 | - | 4, 5 | 544 | |||
AK309431.1 | - | 1, 2 | 99 | |||
AK310359.1 | - | 3, 4 | 415 | |||
AK315424.1 | - | 3, 4 | 412 | |||
AY827074.1 | - | 3, 4 | 467 | |||
BC006250.2 | - | 3, 4 | 461 | 3947694 | ||
BU507838.1 | - | 3, 4 | 525 |