Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRMS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032352.3 | NP_115728.2 | 6, 7 | 742 | ||
XM_005268128.1 | XP_005268185.1 | 6, 7 | 815 | |||
XM_006720274.2 | XP_006720337.1 | 7, 8 | 805 | |||
XM_006720275.2 | XP_006720338.1 | 7, 8 | 805 | |||
XM_017021703.1 | XP_016877192.1 | 6, 7 | 562 | |||
XM_017021704.1 | XP_016877193.1 | 7, 8 | 716 | |||
XM_017021705.1 | XP_016877194.1 | 6, 7 | 639 | |||
GenBank mRNA | AK096496.1 | - | 5, 6 | 465 | ||
AK297106.1 | - | 7, 8 | 805 | |||
AK309431.1 | - | 4, 5 | 360 | |||
AK310359.1 | - | 6, 7 | 676 | |||
AK315424.1 | - | 6, 7 | 673 | |||
BC006250.2 | - | 6, 7 | 722 | 3947694 | ||
BU507838.1 | - | 6, 7 | 786 |