Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321118.1 | NP_001308047.1 | 2 | 614 | ||
NM_001321119.1 | NP_001308048.1 | 2 | 136 | |||
NM_032357.3 | NP_115733.2 | 2 | 435 | |||
NR_104471.1 | 2 | 614 | ||||
NR_135548.1 | 2 | 435 | ||||
GenBank mRNA | AK054693.1 | - | 2 | 401 | ||
AK124405.1 | - | 1 | 257 | |||
AK295922.1 | - | 2 | 271 | |||
BC006429.1 | - | 2 | 197 | 3350642 | ||
BG179982.1 | - | 136 | ||||
DB025106.1 | - | 2 | 436 |