Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167924.1 | NP_001161396.1 | 6, 7 | 519 | ||
NM_032359.3 | NP_115735.2 | 6, 7 | 660 | |||
GenBank mRNA | AK124387.1 | - | 6, 7 | 628 | ||
AK291392.1 | - | 6, 7 | 632 | |||
AK300704.1 | - | 6, 7 | 521 | |||
AK312483.1 | - | 6, 7 | 660 | |||
BC006475.1 | - | 6, 7 | 657 | 2819663 | ||
BC006512.2 | - | 6, 7 | 657 | 2819663 |