Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040107.1 | NP_001035196.1 | 6, 7 | 828 | ||
NM_001256413.1 | NP_001243342.1 | 5, 6 | 788 | |||
NM_032369.3 | NP_115745.2 | 6, 7 | 819 | |||
XM_005253948.2 | XP_005254005.1 | 6, 7 | 906 | |||
XM_011538838.1 | XP_011537140.1 | 7, 8 | 1244 | |||
XM_011538839.2 | XP_011537141.1 | 7, 8 | 978 | |||
XM_011538840.2 | XP_011537142.1 | 7, 8 | 1010 | |||
XM_011538841.2 | XP_011537143.1 | 7, 8 | 988 | |||
XM_011538842.2 | XP_011537144.1 | 7, 8 | 968 | |||
XM_011538844.2 | XP_011537146.1 | 7, 8 | 983 | |||
XM_011538845.1 | XP_011537147.1 | 6, 7 | 863 | |||
XM_011538846.2 | XP_011537148.1 | 7, 8 | 937 | |||
XM_011538847.1 | XP_011537149.1 | 6, 7 | 838 | |||
XM_017020026.1 | XP_016875515.1 | 7, 8 | 952 | |||
XM_017020027.1 | XP_016875516.1 | 6, 7 | 837 | |||
GenBank mRNA | AK293543.1 | - | 5, 6 | 754 | ||
AY359054.1 | - | 6, 7 | 847 | |||
BC007277.1 | - | 6, 7 | 1271 | 3343102 | ||
BC009731.1 | - | 6, 7 | 770 | 3877292 | ||
BC032672.2 | - | 5, 6 | 1392 | 5577070 | ||
GU211002.1 | - | 4, 5 | 638 |