Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040107.1 | NP_001035196.1 | 7 | 915 | ||
NM_001256413.1 | NP_001243342.1 | 6 | 875 | |||
NM_032369.3 | NP_115745.2 | 7 | 906 | |||
XM_005253948.2 | XP_005254005.1 | 7 | 993 | |||
XM_011538838.1 | XP_011537140.1 | 8 | 1331 | |||
XM_011538839.2 | XP_011537141.1 | 8 | 1065 | |||
XM_011538840.2 | XP_011537142.1 | 8 | 1097 | |||
XM_011538841.2 | XP_011537143.1 | 8 | 1075 | |||
XM_011538842.2 | XP_011537144.1 | 8 | 1055 | |||
XM_011538844.2 | XP_011537146.1 | 8 | 1070 | |||
XM_011538845.1 | XP_011537147.1 | 7 | 950 | |||
XM_011538846.2 | XP_011537148.1 | 8 | 1024 | |||
XM_011538847.1 | XP_011537149.1 | 7 | 925 | |||
XM_017020026.1 | XP_016875515.1 | 8 | 1039 | |||
XM_017020027.1 | XP_016875516.1 | 7 | 924 | |||
GenBank mRNA | AK293543.1 | - | 6 | 841 | ||
AY359054.1 | - | 7 | 934 | |||
BC007277.1 | - | 7 | 1358 | 3343102 | ||
BC009731.1 | - | 7 | 857 | 3877292 | ||
BC032672.2 | - | 6 | 1479 | 5577070 | ||
GU211002.1 | - | 5 | 725 |