Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304813.1 | NP_001291742.1 | 5 | 874 | ||
NM_001304814.1 | NP_001291743.1 | 5 | 971 | |||
NM_032376.3 | NP_115752.1 | 4 | 803 | |||
XM_005257737.2 | XP_005257794.1 | 5 | 2151 | |||
XM_011525346.1 | XP_011523648.1 | 7 | 2388 | |||
XM_011525347.1 | XP_011523649.1 | 6 | 2265 | |||
XM_011525349.1 | XP_011523651.1 | 5 | 2171 | |||
XM_011525351.1 | XP_011523653.1 | 5 | 1618 | |||
XM_011525353.1 | XP_011523655.1 | 5 | 2030 | |||
GenBank mRNA | AB097008.1 | - | 4 | 739 | ||
AK130810.1 | - | 4 | 737 | |||
AK172826.1 | - | 4 | 755 | |||
AK294237.1 | - | 4 | 654 | |||
AK313855.1 | - | 4 | 737 | |||
BC007438.2 | - | 4 | 751 | 3028940 |