Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032410.3 | NP_115786.1 | 18 | 1935 | ||
GenBank mRNA | AK090540.1 | - | 8 | 659 | ||
BC048304.1 | - | 18 | 1952 | |||
BC056146.1 | - | 18 | 1816 | 5106726 |