Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318099.1 | NP_001305028.1 | 4 | 1313 | ||
NM_001318100.1 | NP_001305029.1 | 4 | 1390 | |||
NM_001318101.1 | NP_001305030.1 | 2 | 591 | |||
NM_032429.3 | NP_115805.1 | 4 | 1420 | |||
XM_005270222.4 | XP_005270279.1 | 4 | 1652 | |||
XM_017016780.1 | XP_016872269.1 | 3 | 3781 | |||
XM_017016781.1 | XP_016872270.1 | 3 | 761 | |||
GenBank mRNA | AB058716.1 | - | 3 | 4272 | ||
AK097997.1 | - | 3 | 1589 | |||
AK298208.1 | - | 4 | 1278 | |||
AL834338.1 | - | 2 | 796 | |||
AY029201.1 | - | 3 | 1134 | |||
BC005855.2 | - | 1 | 124 | |||
BC006212.2 | - | 4 | 1278 | 3161855 | ||
CX755354.1 | - | 2 | 176 |