Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271286.1 | NP_001258215.1 | 5 | 551 | ||
NM_001271287.1 | NP_001258216.1 | 5 | 579 | |||
NM_001271288.1 | NP_001258217.1 | 5 | 576 | |||
NM_001271289.1 | NP_001258218.1 | 5 | 540 | |||
NM_001271318.1 | NP_001258247.1 | 7 | 739 | |||
NM_032434.3 | NP_115810.2 | 6 | 638 | |||
GenBank mRNA | AB058708.1 | - | 4 | 397 | ||
AK057028.1 | - | 6 | 548 | |||
AK293771.1 | - | 5 | 454 | |||
AK294055.1 | - | 6 | 744 | |||
AK299818.1 | - | 5 | 441 | |||
AK316011.1 | - | 7 | 599 | |||
AK316299.1 | - | 5 | 493 | |||
AL514772.3 | - | 6 | 555 | |||
BC043221.2 | - | 5 | 487 | 5295402 | ||
BC093041.1 | - | 5 | 506 | 6173398 | ||
BG720805.1 | - | 6 | 587 | |||
BX647996.1 | - | 5 | 485 |