Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271286.1 | NP_001258215.1 | 6 | 698 | ||
NM_001271287.1 | NP_001258216.1 | 6 | 726 | |||
NM_001271288.1 | NP_001258217.1 | 6 | 723 | |||
NM_001271289.1 | NP_001258218.1 | 6 | 687 | |||
NM_001271318.1 | NP_001258247.1 | 8 | 886 | |||
NM_032434.3 | NP_115810.2 | 7 | 785 | |||
GenBank mRNA | AB058708.1 | - | 5 | 544 | ||
AK057028.1 | - | 7 | 695 | |||
AK293771.1 | - | 6 | 601 | |||
AK294055.1 | - | 7 | 891 | |||
AK299818.1 | - | 6 | 588 | |||
AK316011.1 | - | 8 | 746 | |||
AK316299.1 | - | 6 | 640 | |||
AL514772.3 | - | 7 | 702 | |||
BC043221.2 | - | 6 | 634 | 5295402 | ||
BC093041.1 | - | 6 | 653 | 6173398 | ||
BX647996.1 | - | 6 | 632 |