Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032435.2 | NP_115811.2 | 5 | 1628 | ||
XM_011544305.2 | XP_011542607.1 | 5 | 1664 | |||
GenBank mRNA | AJ311797.1 | - | 5 | 1628 | ||
AJ311798.1 | - | 5 | 1628 | |||
BC136648.1 | - | 5 | 1487 | 9020638 | ||
BC136649.1 | - | 5 | 1487 | 9020639 |