Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032437.3 | NP_115813.2 | 6 | 1100 | ||
XM_005271274.3 | XP_005271331.1 | 6 | 900 | |||
XM_006710976.3 | XP_006711039.1 | 6 | 994 | |||
XM_006710977.1 | XP_006711040.1 | 6 | 1022 | |||
XM_011542301.2 | XP_011540603.1 | 6 | 1015 | |||
XM_017002548.1 | XP_016858037.1 | 6 | 855 | |||
GenBank mRNA | AB058702.1 | - | 6 | 863 | ||
AK092219.1 | - | 6 | 1011 | |||
AK292220.1 | - | 6 | 1059 | |||
AK315850.1 | - | 6 | 876 | |||
BC015814.1 | - | 6 | 804 | 3915942 |