Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170765.1 | NP_001164236.1 | 8 | 1659 | ||
NM_001170766.1 | NP_001164237.1 | 8 | 1659 | |||
NM_032440.3 | NP_115816.2 | 8 | 1679 | |||
XM_006718033.3 | XP_006718096.1 | 8 | 1967 | |||
XM_017016784.1 | XP_016872273.1 | 7 | 1590 | |||
XM_017016785.1 | XP_016872274.1 | 9 | 1734 | |||
XM_017016786.1 | XP_016872275.1 | 8 | 1655 | |||
XM_017016787.1 | XP_016872276.1 | 7 | 1527 | |||
XM_017016788.1 | XP_016872277.1 | 7 | 1581 | |||
GenBank mRNA | AB058698.1 | - | 8 | 1592 | ||
AK095345.1 | - | 1 | 21 | |||
AL832044.1 | - | 1 | 418 | |||
AL832106.1 | - | 8 | 1639 | |||
AL834245.2 | - | 8 | 1662 | |||
BC053359.1 | - | 8 | 1600 | 6009825 |