Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281520.1 | NP_001268449.1 | 12 | 1785 | ||
NM_001281521.1 | NP_001268450.1 | 12 | 1851 | |||
NM_032485.5 | NP_115874.3 | 12 | 1731 | |||
NM_182802.2 | NP_877954.1 | 12 | 1737 | |||
XM_011529387.2 | XP_011527689.1 | 12 | 1694 | |||
XM_017028105.1 | XP_016883594.1 | 12 | 1880 | |||
XM_017028106.1 | XP_016883595.1 | 12 | 1697 | |||
XM_017028107.1 | XP_016883596.1 | 5 | 1970 | |||
GenBank mRNA | AJ439063.1 | - | 12 | 1731 | ||
AK027644.1 | - | 6 | 770 | |||
AK314654.1 | - | 12 | 1413 | |||
AY158211.1 | - | 12 | 1436 | |||
BC005170.1 | - | 12 | 1436 | |||
BC008830.2 | - | 9 | 1041 | |||
BC080656.1 | - | 12 | 1436 | 4841271 | ||
BC101055.1 | - | 12 | 1652 | 40008095 | ||
BC101056.1 | - | 12 | 1532 | 40008096 | ||
BC101057.1 | - | 12 | 1586 | 40008097 | ||
KU178803.1 | - | 6 | 643 | |||
KU178804.1 | - | 6 | 643 |