Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281520.1 | NP_001268449.1 | 16, 17 | 2602 | ||
NM_001281521.1 | NP_001268450.1 | 16, 17 | 2668 | |||
NM_001281522.1 | NP_001268451.1 | 15, 16 | 2407 | |||
NM_032485.5 | NP_115874.3 | 16, 17 | 2548 | |||
NM_182802.2 | NP_877954.1 | 16, 17 | 2554 | |||
XM_017028105.1 | XP_016883594.1 | 16, 17 | 2697 | |||
XM_017028106.1 | XP_016883595.1 | 16, 17 | 2514 | |||
XM_017028107.1 | XP_016883596.1 | 9, 10 | 2787 | |||
GenBank mRNA | AJ439063.1 | - | 16, 17 | 2548 | ||
AK027644.1 | - | 10, 11 | 1587 | |||
AK314654.1 | - | 16, 17 | 2230 | |||
AY158211.1 | - | 16, 17 | 2253 | |||
BC005170.1 | - | 16, 17 | 2253 | |||
BC008830.2 | - | 13, 14 | 1858 | |||
BC080656.1 | - | 16, 17 | 2253 | 4841271 | ||
BC101054.1 | - | 15, 16 | 2208 | 40008094 | ||
BC101055.1 | - | 16, 17 | 2469 | 40008095 | ||
BC101056.1 | - | 16, 17 | 2349 | 40008096 | ||
BC101057.1 | - | 16, 17 | 2403 | 40008097 | ||
KU178803.1 | - | 10, 11 | 1460 | |||
KU178804.1 | - | 11, 12 | 1551 | |||
KU178805.1 | - | 9, 10 | 1319 |