Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032489.2 | NP_115878.2 | 10 | 1651 | ||
GenBank mRNA | AB051833.1 | - | 10 | 1651 | ||
AF085884.1 | - | 2 | 271 | |||
AK302290.1 | - | 10 | 1549 | |||
AK311572.1 | - | 10 | 1458 | |||
AK312782.1 | - | 10 | 1669 | |||
BC033010.2 | - | 10 | 1648 | 4822765 |