Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032494.2 | NP_115883.2 | 3 | 406 | ||
XM_011511998.1 | XP_011510300.1 | 3 | 351 | |||
XM_017005107.1 | XP_016860596.1 | 3 | 359 | |||
GenBank mRNA | AF334161.1 | - | 3 | 392 | ||
AK315329.1 | - | 3 | 360 | |||
BC032001.1 | - | 3 | 344 | 4749794 | ||
BG773518.1 | - | 3 | 435 |