Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194286.3 | NP_919262.2 | 6 | 886 | ||
GenBank mRNA | AB058756.1 | - | 6 | 790 | ||
AK291361.1 | - | 6 | 886 | |||
BC152471.1 | - | 6 | 801 | 8862608 | ||
BC167162.1 | - | 6 | 790 | 8862617 | ||
BC167163.1 | - | 6 | 790 | 8862618 | ||
KC995185.1 | - | 6 | 498 | |||
KC995186.1 | - | 6 | 498 |