Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032505.2 | NP_115894.2 | 3 | 524 | ||
XM_005264771.3 | XP_005264828.1 | 2 | 309 | |||
GenBank mRNA | AB058745.1 | - | 1 | 247 | ||
AF385438.1 | - | 2 | 457 | |||
AK096640.1 | - | 3 | 520 | |||
BC117487.1 | - | 2 | 421 | 40126038 | ||
DA246470.1 | - | 3 | 520 | |||
DB065239.1 | - | 3 | 525 |