Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032509.3 | NP_115898.2 | 5 | 355 | ||
GenBank mRNA | AF251062.1 | - | 5 | 330 | ||
AK027469.1 | - | 5 | 340 | |||
AK225476.1 | - | 5 | 341 | |||
AK314491.1 | - | 5 | 343 | |||
BC015965.1 | - | 5 | 340 | |||
BC028230.1 | - | 5 | 323 | 5240795 | ||
BC039740.1 | - | 5 | 336 | 6066906 | ||
BC050528.1 | - | 5 | 323 | 6471474 | ||
BP281245.1 | - | 5 | 356 |