Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPTG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032520.4 | NP_115909.1 | 5, 6 | 375 | ||
XM_017023782.1 | XP_016879271.1 | 5, 6 | 391 | |||
XM_017023783.1 | XP_016879272.1 | 3, 4 | 263 | |||
GenBank mRNA | AK126110.1 | - | 1, 2 | 1324 | ||
AK312067.1 | - | 5, 6 | 343 | |||
AY203933.1 | - | 5, 6 | 306 | |||
BC014592.1 | - | 5, 6 | 338 | 4824054 | ||
BC131525.1 | - | 4, 5 | 270 | |||
BU145878.1 | - | 5, 6 | 351 | |||
DT219524.1 | - | 5, 6 | 376 |