Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPTG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032520.4 | NP_115909.1 | 6 | 427 | ||
XM_017023782.1 | XP_016879271.1 | 6 | 443 | |||
XM_017023783.1 | XP_016879272.1 | 4 | 315 | |||
GenBank mRNA | AF302786.1 | - | 6 | 360 | ||
AK126110.1 | - | 2 | 1376 | |||
AK312067.1 | - | 6 | 395 | |||
AY203933.1 | - | 6 | 358 | |||
BC014592.1 | - | 6 | 390 | 4824054 | ||
BC131525.1 | - | 5 | 322 | |||
BU145878.1 | - | 6 | 403 | |||
BX398427.1 | - | 6 | 804 | |||
DT219524.1 | - | 6 | 428 |