Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032536.2 | NP_115925.2 | 6 | 1983 | ||
XM_006717304.3 | XP_006717367.1 | 9 | 2106 | |||
XM_011519094.2 | XP_011517396.1 | 11 | 2322 | |||
XM_011519096.2 | XP_011517398.1 | 11 | 2719 | |||
XM_011519097.2 | XP_011517399.1 | 10 | 2240 | |||
XM_011519098.2 | XP_011517400.1 | 11 | 2272 | |||
XM_011519099.2 | XP_011517401.1 | 10 | 2243 | |||
XM_011519100.2 | XP_011517402.1 | 9 | 2172 | |||
XM_011519102.2 | XP_011517404.1 | 10 | 2187 | |||
XM_011519103.2 | XP_011517405.1 | 9 | 2079 | |||
XM_011519104.2 | XP_011517406.1 | 9 | 2303 | |||
XM_011519105.2 | XP_011517407.1 | 9 | 2169 | |||
XM_011519106.2 | XP_011517408.1 | 10 | 2253 | |||
XM_011519107.2 | XP_011517409.1 | 8 | 2004 | |||
XM_011519112.2 | XP_011517414.1 | 4 | 1291 | |||
XM_011519113.2 | XP_011517415.1 | 3 | 1222 | |||
XM_017015212.1 | XP_016870701.1 | 12 | 2426 | |||
XM_017015213.1 | XP_016870702.1 | 10 | 2629 | |||
XM_017015214.1 | XP_016870703.1 | 9 | 2151 | |||
GenBank mRNA | AB058760.2 | - | 6 | 1983 | ||
AF131842.1 | - | 4 | 397 | |||
BC013770.1 | - | 5 | 1292 | 4344617 |