Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144003.2 | NP_001137475.1 | 5 | 1855 | ||
NM_001144004.2 | NP_001137476.1 | 5 | 1843 | |||
NM_001144005.2 | NP_001137477.1 | 5 | 1865 | |||
NM_001144006.2 | NP_001137478.1 | 3 | 2075 | |||
NM_001144008.2 | NP_001137480.1 | 3 | 2087 | |||
NM_001144009.2 | NP_001137481.1 | 2 | 1928 | |||
NM_001144010.2 | NP_001137482.1 | 3 | 2065 | |||
NM_032539.4 | NP_115928.1 | 5 | 2338 | |||
XM_005262342.3 | XP_005262399.1 | 5 | 2940 | |||
XM_005262343.3 | XP_005262400.1 | 5 | 2950 | |||
XM_005262344.3 | XP_005262401.1 | 5 | 2928 | |||
XM_005262345.3 | XP_005262402.1 | 4 | 2791 | |||
GenBank mRNA | AB058757.1 | - | 1 | 5603 | ||
AK091015.1 | - | 1 | 1456 | |||
AK096278.1 | - | 5 | 2334 | |||
AK289732.1 | - | 5 | 1705 | |||
BC113011.2 | - | 3 | 1929 | 40028691 | ||
BC113012.2 | - | 3 | 1931 | 40028693 | ||
CR627412.1 | - | 2 | 3377 |