Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290325.1 | NP_001277254.1 | 9 | 3026 | ||
NM_001290326.1 | NP_001277255.1 | 10 | 3142 | |||
NM_032557.6 | NP_115946.2 | 9 | 3026 | |||
XM_011532360.2 | XP_011530662.1 | 8 | 2412 | |||
GenBank mRNA | AB067478.1 | - | 8 | 1707 | ||
AF211481.1 | - | 1 | 599 | |||
AK057992.1 | - | 6 | 1550 | |||
AK091712.1 | - | 1 | 896 | |||
AK093289.1 | - | 1 | 76 | |||
AK093754.1 | - | 10 | 2365 | |||
AK126943.1 | - | 10 | 2854 | |||
AL833976.1 | - | 9 | 2998 | |||
BC017668.2 | - | 2 | 1663 | |||
BC039115.1 | - | 9 | 3026 | 5258804 | ||
BC068975.1 | - | 9 | 2973 | 6067501 | ||
BG575871.1 | - | 1 | 135 |