Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032566.2 | NP_115955.1 | 3 | 213 | ||
GenBank mRNA | AF268198.1 | - | 3 | 213 | ||
AK311570.1 | - | 2 | 914 | |||
AY359072.1 | - | 3 | 200 | |||
BC109385.1 | - | 3 | 176 | 40013896 | ||
BC110067.1 | - | 3 | 176 | 40013897 |