Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308096.1 | NP_001295025.1 | 5 | 411 | ||
NM_001324098.1 | NP_001311027.1 | 5 | 497 | |||
NM_032569.3 | NP_115958.2 | 5 | 411 | |||
NR_136695.1 | 6 | 696 | ||||
NR_136696.1 | 5 | 497 | ||||
NR_136697.1 | 5 | 497 | ||||
NR_136698.1 | 5 | 497 | ||||
NR_136699.1 | 5 | 497 | ||||
NR_136700.1 | 5 | 497 | ||||
XM_011522716.2 | XP_011521018.1 | 5 | 539 | |||
XM_011522717.1 | XP_011521019.1 | 6 | 596 | |||
XM_017023786.1 | XP_016879275.1 | 5 | 492 | |||
XM_017023787.1 | XP_016879276.1 | 5 | 492 | |||
XM_017023788.1 | XP_016879277.1 | 6 | 596 | |||
XM_017023789.1 | XP_016879278.1 | 6 | 596 | |||
GenBank mRNA | AF244907.1 | - | 5 | 392 | ||
AF326966.1 | - | 5 | 374 | |||
AK307834.1 | - | 5 | 387 | |||
AY352585.1 | - | 5 | 374 | |||
BC003693.1 | - | 4 | 336 | |||
BC032855.1 | - | 5 | 402 | 5265561 | ||
BC047223.1 | - | 5 | 391 | 5263991 | ||
BC064940.1 | - | 6 | 595 | 5787345 | ||
DB492328.1 | - | 5 | 416 |