Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032582.3 | NP_115971.2 | 13 | 1638 | ||
XM_011525371.1 | XP_011523673.1 | 13 | 1519 | |||
XM_011525372.1 | XP_011523674.1 | 13 | 1519 | |||
XM_011525373.1 | XP_011523675.1 | 13 | 1547 | |||
XM_011525374.1 | XP_011523676.1 | 13 | 1364 | |||
XM_011525375.1 | XP_011523677.1 | 13 | 1337 | |||
XM_011525376.1 | XP_011523678.1 | 13 | 1364 | |||
XM_011525378.1 | XP_011523680.1 | 12 | 1229 | |||
XM_011525379.2 | XP_011523681.1 | 9 | 852 | |||
XM_017025233.1 | XP_016880722.1 | 12 | 1303 | |||
XM_017025234.1 | XP_016880723.1 | 5 | 399 | |||
GenBank mRNA | AF350251.1 | - | 5 | 391 | ||
AF533230.1 | - | 13 | 1524 | |||
AK057301.1 | - | 13 | 1364 |