Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032595.4 | NP_115984.3 | 6 | 1940 | ||
GenBank mRNA | AJ401189.1 | - | 6 | 1936 | ||
AL713642.1 | - | 6 | 747 | |||
BC016162.1 | - | 4 | 427 |