Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282788.1 | NP_001269717.1 | 6, 7 | 1115 | ||
NM_001282789.1 | NP_001269718.1 | 7, 8 | 1062 | |||
NM_032599.3 | NP_115988.1 | 6, 7 | 1121 | |||
NR_104242.1 | 7, 8 | 1419 | ||||
NR_104243.1 | 8, 9 | 1308 | ||||
GenBank mRNA | AF367470.1 | - | 6, 7 | 1074 | ||
AK093161.1 | - | 7, 8 | 1059 | |||
AK313608.1 | - | 1066 | ||||
BC037248.1 | - | 6, 7 | 1054 | 5165912 |