Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032623.3 | NP_116012.2 | 3 | 400 | ||
GenBank mRNA | AF329088.1 | - | 3 | 287 | ||
AF484960.1 | - | 3 | 400 | |||
BC104173.1 | - | 3 | 286 | 40030666 | ||
BC104174.1 | - | 3 | 286 | 40030667 |